siRNA Details
Gene Symbol: ZNF517 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317936.1 | NP_001304865.1 | 5 | 520 | ||
NM_213605.2 | NP_998770.2 | 5 | 541 | |||
NR_134284.1 | 5 | 541 | ||||
XM_011517014.2 | XP_011515316.1 | 5 | 1080 | |||
XM_011517015.2 | XP_011515317.1 | 5 | 522 | |||
XM_011517017.2 | XP_011515319.1 | 4 | 939 | |||
XM_017013384.1 | XP_016868873.1 | 2 | 465 | |||
GenBank mRNA | AK096527.1 | - | 6 | 1037 | ||
AK097278.1 | - | 3 | 1564 | |||
AK131440.1 | - | 5 | 541 | |||
BC126161.1 | - | 4 | 461 | 8991877 | ||
BC143922.1 | - | 3 | 334 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
