siRNA Details
Gene Symbol: APH1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077628.2 | NP_001071096.1 | 1 | 436 | ||
NM_001243771.1 | NP_001230700.1 | 1 | 436 | |||
NM_001243772.1 | NP_001230701.1 | 1 | 436 | |||
NM_016022.3 | NP_057106.2 | 1 | 436 | |||
NR_045033.1 | 1 | 436 | ||||
NR_045034.1 | 1 | 436 | ||||
NR_045035.1 | 1 | 436 | ||||
XM_017001417.1 | XP_016856906.1 | 3 | 1019 | |||
GenBank mRNA | AF151835.1 | - | 1 | 138 | ||
AF508787.1 | - | 1 | 94 | |||
AK027879.1 | - | 1 | 206 | |||
AK075295.1 | - | 1 | 155 | |||
AK075452.1 | - | 1 | 244 | |||
AK297680.1 | - | 1 | 202 | |||
AK298832.1 | - | 1 | 243 | |||
AK298876.1 | - | 1 | 215 | |||
AK300640.1 | - | 1 | 241 | |||
AK307358.1 | - | 1 | 243 | |||
AK314490.1 | - | 1 | 206 | |||
AX775955.1 | - | 1 | 172 | |||
AX775957.1 | - | 1 | 226 | |||
AY113698.1 | - | 1 | 94 | |||
AY113699.1 | - | 1 | 94 | |||
AY358951.1 | - | 1 | 240 | |||
BC001230.2 | - | 1 | 206 | 3457593 | ||
BC008732.2 | - | 1 | 193 | 3162330 | ||
BC009501.2 | - | 1 | 227 | 4040230 | ||
BC015568.2 | - | 1 | 195 | 3356401 | ||
BC017699.1 | - | 1 | 172 | 4177580 | ||
BC020590.1 | - | 1 | 226 | 4420942 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
