siRNA Details
Gene Symbol: UAP1L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207309.2 | NP_997192.2 | 3 | 657 | ||
XM_006717317.3 | XP_006717380.1 | 3 | 683 | |||
XM_011519182.2 | XP_011517484.1 | 2 | 251 | |||
GenBank mRNA | AK074143.1 | - | 1 | 34 | ||
AK095393.1 | - | 3 | 398 | |||
AK128529.1 | - | 1 | 492 | |||
AL832421.1 | - | 3 | 426 | |||
BC106035.1 | - | 3 | 369 | |||
BC136391.1 | - | 2 | 224 | |||
BX333979.2 | - | 3 | 656 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
