siRNA Details
Gene Symbol: UAP1L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207309.2 | NP_997192.2 | 9 | 1746 | ||
GenBank mRNA | AK022632.1 | - | 4 | 631 | ||
AK074143.1 | - | 4 | 2794 | |||
AK095393.1 | - | 9 | 1487 | |||
AK128529.1 | - | 7 | 1581 | |||
AL832421.1 | - | 9 | 1515 | |||
BC106035.1 | - | 9 | 1458 | |||
BC136391.1 | - | 8 | 1313 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
