siRNA Details
Gene Symbol: TRIM58 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BX537432.1 | - | 3 | 357 |
Gene Symbol: OR2W3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001957.2 | NP_001001957.2 | 1 | 100 | ||
GenBank mRNA | BC146848.1 | - | 1 | 131 | 9056776 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
