siRNA Details
Gene Symbol: LINC01105 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026832.1 | 2 | 4391 | |||
GenBank mRNA | AK055156.1 | - | 1 | 1162 | ||
AK091363.1 | - | 1 | 1784 | |||
AL713643.1 | - | 1 | 1769 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
