siRNA Details
Gene Symbol: NKIRAS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020345.3 | NP_065078.1 | 5 | 867 | ||
XM_005265076.3 | XP_005265133.1 | 5 | 754 | |||
XM_005265077.4 | XP_005265134.1 | 4 | 722 | |||
XM_005265078.3 | XP_005265135.1 | 5 | 706 | |||
GenBank mRNA | AF229839.1 | - | 3 | 465 | ||
AK027749.1 | - | 5 | 844 | |||
BC012145.1 | - | 5 | 750 | 4549097 | ||
BC066940.1 | - | 5 | 758 | 5273965 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
