siRNA Details
Gene Symbol: TRDN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001251987.1 | NP_001238916.1 | 10 | 1228 | ||
NM_001256020.1 | NP_001242949.1 | 9 | 1168 | |||
NM_006073.3 | NP_006064.2 | 10 | 1228 | |||
GenBank mRNA | AJ489257.2 | - | 10 | 1123 | ||
AK291867.1 | - | 10 | 1085 | |||
AK307619.1 | - | 10 | 905 | |||
BC029393.1 | - | 10 | 1085 | |||
BC057222.1 | - | 10 | 1089 | |||
BC139910.1 | - | 9 | 984 | |||
U18985.1 | - | 10 | 932 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
