siRNA Details
Gene Symbol: TXNDC12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_046405.1 | 3 | 1564 | |||
NR_046406.1 | 3 | 1441 |
Gene Symbol: KTI12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138417.2 | NP_612426.1 | 1 | 80 | ||
GenBank mRNA | AF327348.1 | - | 1 | 61 | ||
AK313746.1 | - | 1 | 124 | |||
BC012173.1 | - | 1 | 44 | 4643409 | ||
BI909162.1 | - | 1 | 96 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
