siRNA Details
Gene Symbol: CTNNA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164883.1 | NP_001158355.1 | 14 | 2210 | ||
NM_001282597.2 | NP_001269526.1 | 14 | 2210 | |||
NM_001282598.1 | NP_001269527.1 | 14 | 2063 | |||
NM_001282599.1 | NP_001269528.1 | 8 | 1151 | |||
NM_001282600.1 | NP_001269529.1 | 8 | 1094 | |||
NM_001320810.1 | NP_001307739.1 | 8 | 1094 | |||
NM_004389.3 | NP_004380.2 | 14 | 2210 | |||
XM_011532555.2 | XP_011530857.1 | 14 | 2057 | |||
XM_011532556.2 | XP_011530858.1 | 15 | 2253 | |||
XM_011532557.2 | XP_011530859.1 | 6 | 799 | |||
XM_017003403.1 | XP_016858892.1 | 15 | 2253 | |||
XM_017003404.1 | XP_016858893.1 | 14 | 2148 | |||
XM_017003405.1 | XP_016858894.1 | 14 | 2148 | |||
XM_017003406.1 | XP_016858895.1 | 8 | 1184 | |||
GenBank mRNA | AK127226.1 | - | 18 | 2656 | ||
AK295181.1 | - | 14 | 2009 | |||
AK295493.1 | - | 8 | 1081 | |||
AK303035.1 | - | 14 | 2063 | |||
BC040458.1 | - | 13 | 1882 | 4815068 | ||
BX537769.1 | - | 8 | 1151 | |||
BX648368.1 | - | 1 | 23 | |||
HQ589336.1 | - | 8 | 1082 | |||
M94151.2 | - | 13 | 1961 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
