siRNA Details
Gene Symbol: CTNNA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164883.1 | NP_001158355.1 | 17 | 2834 | ||
NM_001282597.2 | NP_001269526.1 | 19 | 3113 | |||
NM_001282598.1 | NP_001269527.1 | 18 | 2822 | |||
NM_001282599.1 | NP_001269528.1 | 12 | 1910 | |||
NM_001282600.1 | NP_001269529.1 | 12 | 1853 | |||
NM_001320810.1 | NP_001307739.1 | 13 | 1997 | |||
NM_004389.3 | NP_004380.2 | 18 | 2969 | |||
XM_011532555.2 | XP_011530857.1 | 19 | 2960 | |||
XM_011532556.2 | XP_011530858.1 | 20 | 3156 | |||
XM_011532557.2 | XP_011530859.1 | 11 | 1702 | |||
XM_017003403.1 | XP_016858892.1 | 19 | 3012 | |||
XM_017003406.1 | XP_016858895.1 | 11 | 1808 | |||
GenBank mRNA | AK127226.1 | - | 22 | 3415 | ||
AK294362.1 | - | 4 | 934 | |||
AK295181.1 | - | 17 | 2633 | |||
AK295493.1 | - | 12 | 1840 | |||
AK303035.1 | - | 18 | 2822 | |||
BC040458.1 | - | 17 | 2641 | 4815068 | ||
BC052996.1 | - | 18 | 2903 | 6187158 | ||
BX537769.1 | - | 12 | 1910 | |||
BX648368.1 | - | 5 | 782 | |||
HQ589336.1 | - | 12 | 1841 | |||
M94151.2 | - | 18 | 2864 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
