siRNA Details
Gene Symbol: LRRTM3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178011.4 | NP_821079.3 | 2 | 914 | ||
GenBank mRNA | AK290168.1 | - | 2 | 447 | ||
AY182028.1 | - | 1 | 364 | |||
AY358315.1 | - | 2 | 871 | |||
BC111492.1 | - | 2 | 866 | 5196989 | ||
BC113715.1 | - | 2 | 424 | 8322767 | ||
BC113717.1 | - | 2 | 424 | 8322769 | ||
BX640611.1 | - | 2 | 893 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
