siRNA Details
Gene Symbol: SLC9A8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001260491.1 | NP_001247420.1 | 16 | 5980 | ||
NM_015266.2 | NP_056081.1 | 16 | 5932 | |||
NR_048537.1 | 15 | 5897 | ||||
NR_048538.1 | 14 | 5813 | ||||
NR_048539.1 | 14 | 5769 | ||||
NR_048540.1 | 12 | 5595 | ||||
XM_006723756.1 | XP_006723819.1 | 17 | 5893 | |||
XM_011528736.1 | XP_011527038.1 | 17 | 5941 | |||
XM_011528737.1 | XP_011527039.1 | 16 | 5814 | |||
XM_011528738.1 | XP_011527040.1 | 16 | 5824 | |||
XM_011528739.1 | XP_011527041.1 | 16 | 5818 | |||
XM_011528741.1 | XP_011527043.1 | 15 | 5725 | |||
XM_011528742.1 | XP_011527044.1 | 15 | 5714 | |||
XM_011528743.2 | XP_011527045.1 | 9 | 5078 | |||
XM_011528744.2 | XP_011527046.1 | 8 | 4909 | |||
XM_017027755.1 | XP_016883244.1 | 16 | 5802 | |||
XM_017027756.1 | XP_016883245.1 | 17 | 5890 | |||
GenBank mRNA | AB023156.2 | - | 16 | 5764 | ||
AK124491.1 | - | 3 | 2145 | |||
AL049469.1 | - | 1 | 2001 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
