siRNA Details
Gene Symbol: RPS18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022551.2 | NP_072045.1 | 1 | 19 | ||
GenBank mRNA | AB055778.1 | - | 1 | 17 | ||
BC101786.1 | - | 1 | 14 | 8069292 | ||
BC101788.1 | - | 1 | 14 | 8069294 | ||
BC106063.1 | - | 1 | 72 | 4386992 | ||
X69150.1 | - | 1 | 20 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
