siRNA Details
Gene Symbol: HSPE1-MOB4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001202485.1 | NP_001189414.1 | 1 | 419 | ||
GenBank mRNA | BJ992310.1 | - | 1 | 37 |
Gene Symbol: HSPE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002157.2 | NP_002148.1 | 1 | 419 | ||
GenBank mRNA | AK312104.1 | - | 1 | 15 | ||
BC023518.2 | - | 1 | 93 | 4304224 | ||
CN264788.1 | - | 1 | 424 | |||
U07550.1 | - | 1 | 20 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
