siRNA Details
Gene Symbol: SRSF3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003017.4 | NP_003008.1 | 1 | 158 | ||
NR_036610.1 | 1 | 158 | ||||
GenBank mRNA | AF107405.1 | - | 1 | 82 | ||
AK304097.1 | - | 1 | 105 | |||
AK312553.1 | - | 1 | 109 | |||
BC000914.1 | - | 1 | 92 | 3049167 | ||
BC069018.1 | - | 1 | 88 | 3507371 | ||
D28423.1 | - | 1 | 105 | |||
DC398250.1 | - | 1 | 250 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
