siRNA Details
Gene Symbol: CHD1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256336.1 | NP_001243265.1 | 1 | 170 | ||
NM_001256337.1 | NP_001243266.1 | 1 | 170 | |||
NM_001256338.1 | NP_001243267.1 | 1 | 170 | |||
NM_004284.4 | NP_004275.4 | 1 | 170 | |||
NM_024568.2 | NP_078844.2 | 1 | 170 | |||
NR_046070.1 | 1 | 170 | ||||
XM_006711639.3 | XP_006711702.1 | 1 | 151 | |||
XM_017002860.1 | XP_016858349.1 | 1 | 118 | |||
XM_017002861.1 | XP_016858350.1 | 1 | 118 | |||
XM_017002863.1 | XP_016858352.1 | 1 | 46 | |||
XM_017002864.1 | XP_016858353.1 | 1 | 118 | |||
GenBank mRNA | AF537213.1 | - | 1 | 127 | ||
AK001342.1 | - | 1 | 127 | |||
AK027631.1 | - | 1 | 127 | |||
AK293157.1 | - | 1 | 105 | |||
AK297135.1 | - | 1 | 108 | |||
AK298424.1 | - | 1 | 110 | |||
AK307400.1 | - | 1 | 127 | |||
BC001171.1 | - | 1 | 104 | |||
BC005038.1 | - | 1 | 67 | |||
BC043501.1 | - | 1 | 88 | |||
BC077717.1 | - | 1 | 107 | 6573666 | ||
DA007597.1 | - | 1 | 148 | |||
DA155685.1 | - | 1 | 171 | |||
EF560738.1 | - | 1 | 129 | |||
KJ534809.1 | - | 1 | 107 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
