siRNA Details
Gene Symbol: MAVS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001206491.1 | NP_001193420.1 | 1, 2 | 113 | ||
NM_020746.4 | NP_065797.2 | 1, 2 | 113 | |||
NR_037921.1 | 1, 2 | 113 | ||||
GenBank mRNA | AB033097.1 | - | 1 | 19 | ||
AB097003.1 | - | 1, 2 | 76 | |||
AB232371.1 | - | 1 | 13 | |||
AK023799.1 | - | 1, 2 | 69 | |||
AK123956.1 | - | 1, 2 | 74 | |||
AK130992.1 | - | 1, 2 | 34 | |||
AK291785.1 | - | 1, 2 | 72 | |||
AK296897.1 | - | 1, 2 | 78 | |||
BC044952.1 | - | 1, 2 | 47 | 5751684 | ||
BM834250.1 | - | 1, 2 | 77 | |||
BX649084.1 | - | 1, 2 | 74 | |||
DB219666.1 | - | 1, 2 | 113 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
