siRNA Details
Gene Symbol: AHI1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134830.1 | NP_001128302.1 | 1 | 100 | ||
NM_001134831.1 | NP_001128303.1 | 1 | 100 | |||
NM_001134832.1 | NP_001128304.1 | 1 | 100 | |||
NM_017651.4 | NP_060121.3 | 1 | 100 | |||
XM_011535910.2 | XP_011534212.1 | 1 | 73 | |||
XM_011535911.2 | XP_011534213.1 | 1 | 36 | |||
XM_017010977.1 | XP_016866466.1 | 1 | 110 | |||
XM_017010981.1 | XP_016866470.1 | 1 | 106 | |||
GenBank mRNA | AJ459824.1 | - | 1 | 80 | ||
AJ459825.1 | - | 1 | 80 | |||
AJ606362.1 | - | 1 | 80 | |||
AK092262.1 | - | 1 | 73 | |||
BC094800.1 | - | 1 | 75 | 4823262 | ||
BX643029.1 | - | 1 | 101 | |||
DQ090887.1 | - | 1 | 54 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
