siRNA Details
Gene Symbol: ZNF160 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001102603.1 | NP_001096073.1 | 1 | 45 | ||
NM_001322125.1 | NP_001309054.1 | 1 | 45 | |||
NM_001322126.1 | NP_001309055.1 | 1 | 45 | |||
NM_001322128.1 | NP_001309057.1 | 1 | 45 | |||
NM_001322129.1 | NP_001309058.1 | 1 | 45 | |||
NM_001322130.1 | NP_001309059.1 | 1 | 45 | |||
NM_001322131.1 | NP_001309060.1 | 1 | 45 | |||
NM_001322132.1 | NP_001309061.1 | 1 | 45 | |||
NM_001322133.1 | NP_001309062.1 | 1 | 45 | |||
NM_001322134.1 | NP_001309063.1 | 1 | 45 | |||
NM_001322135.1 | NP_001309064.1 | 1 | 45 | |||
NM_001322137.1 | NP_001309066.1 | 1 | 45 | |||
NM_001322138.1 | NP_001309067.1 | 1 | 45 | |||
NM_001322139.1 | NP_001309068.1 | 1 | 45 | |||
NM_033288.3 | NP_150630.2 | 1 | 45 | |||
NM_198893.2 | NP_942596.1 | 1 | 45 | |||
GenBank mRNA | BC000807.1 | - | 1 | 14 | ||
DA230397.1 | - | 1 | 107 | |||
DB078640.1 | - | 1 | 45 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
