siRNA Details
Gene Symbol: ZNF787 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001002836.3 | NP_001002836.2 | 1 | 175 | ||
GenBank mRNA | BC012495.1 | - | 1 | 79 | ||
BC077728.1 | - | 1 | 82 | 6210543 | ||
HY059733.1 | - | 1 | 176 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
