siRNA Details
Gene Symbol: SRP9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130440.1 | NP_001123912.1 | 1 | 46 | ||
NM_003133.5 | NP_003124.1 | 1 | 46 | |||
GenBank mRNA | AF070649.1 | - | 1 | 25 | ||
AK289595.1 | - | 11 | ||||
BC008443.1 | - | 1 | 30 | 4102222 | ||
BC015094.1 | - | 1 | 17 | 3930047 | ||
BC021995.1 | - | 1 | 26 | 4101294 | ||
BC066957.1 | - | 1 | 33 | 5263635 | ||
BC106005.1 | - | 1 | 22 | 6026221 | ||
DA983529.1 | - | 1 | 46 | |||
DB194289.1 | - | 1 | 51 | |||
EF488978.1 | - | 1 | 30 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
