siRNA Details
Gene Symbol: DAAM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001201427.1 | NP_001188356.1 | 2 | 99 | ||
NM_015345.3 | NP_056160.2 | 2 | 163 | |||
XM_006715039.3 | XP_006715102.3 | 2 | 744 | |||
XM_006715040.3 | XP_006715103.3 | 2 | 743 | |||
XM_006715042.2 | XP_006715105.1 | 2 | 2739 | |||
XM_006715043.2 | XP_006715106.1 | 2 | 116 | |||
XM_006715045.3 | XP_006715108.1 | 2 | 1331 | |||
XM_006715046.3 | XP_006715109.1 | 2 | 509 | |||
XM_017010630.1 | XP_016866119.1 | 2 | 1355 | |||
GenBank mRNA | AB002379.2 | - | 2 | 99 | ||
AK310941.1 | - | 2 | 106 | |||
AL596701.1 | - | 2 | 189 | |||
AL833083.2 | - | 2 | 143 | |||
BC078153.1 | - | 2 | 388 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
