siRNA Details
Gene Symbol: AGBL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178563.3 | NP_848658.3 | 1 | 133 | ||
XM_017012136.1 | XP_016867625.1 | 1 | 1469 | |||
XM_017012137.1 | XP_016867626.1 | 1 | 1469 | |||
XM_017012141.1 | XP_016867630.1 | 1 | 1472 | |||
GenBank mRNA | AK023045.1 | - | 1 | 85 | ||
AK302827.1 | - | 1 | 133 | |||
AK311252.1 | - | 1 | 74 | |||
AL832281.1 | - | 1 | 113 | |||
BC030651.2 | - | 1 | 122 | 5267615 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
