siRNA Details
Gene Symbol: DISP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032890.3 | NP_116279.2 | 3 | 198 | ||
XM_005273335.2 | XP_005273392.1 | 1 | 39 | |||
XM_006711592.2 | XP_006711655.1 | 1 | 905 | |||
GenBank mRNA | AK125728.1 | - | 1 | 36 | ||
BC007734.2 | - | 1 | 19 | |||
BC107762.1 | - | 1 | 23 | |||
DA756409.1 | - | 3 | 198 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
