siRNA Details
Gene Symbol: RIBC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001031745.4 | NP_001026915.1 | 2 | 202 | ||
NM_001267053.3 | NP_001253982.1 | 2 | 202 | |||
NM_144968.3 | NP_659405.1 | 2 | 202 | |||
XM_005261988.3 | XP_005262045.1 | 2 | 410 | |||
XM_005261990.3 | XP_005262047.1 | 2 | 276 | |||
GenBank mRNA | AK057345.1 | - | 2 | 168 | ||
AK290346.1 | - | 2 | 152 | |||
AK292871.1 | - | 2 | 157 | |||
AK304179.1 | - | 2 | 202 | |||
BC036767.1 | - | 2 | 117 | 5744127 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
