siRNA Details
Gene Symbol: RPL26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000987.3 | NP_000978.1 | 1 | 82 | ||
GenBank mRNA | AK311804.1 | - | 1 | 28 | ||
BC071664.1 | - | 1 | 17 | 5189436 | ||
BI091167.1 | - | 1 | 82 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
