siRNA Details
Gene Symbol: RPL29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000992.2 | NP_000983.1 | 1 | 24 | ||
GenBank mRNA | BC008926.2 | - | 1 | 24 | 3835176 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
