siRNA Details
Gene Symbol: C4orf36 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144645.3 | NP_653246.1 | 1 | 144 | ||
XM_011531611.2 | XP_011529913.1 | 1 | 133 | |||
GenBank mRNA | BC016746.1 | - | 1 | 120 | 4827131 | |
BP289794.1 | - | 1 | 144 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
