siRNA Details
Gene Symbol: LARP7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001267039.1 | NP_001253968.1 | 6, 7 | 873 | ||
NM_015454.2 | NP_056269.1 | 4, 5 | 537 | |||
NM_016648.3 | NP_057732.2 | 4, 5 | 665 | |||
NR_049768.1 | 4, 5 | 562 | ||||
GenBank mRNA | AF068284.1 | - | 2, 3 | 112 | ||
AK000274.1 | - | 4, 5 | 475 | |||
AK225900.1 | - | 1, 2 | 34 | |||
BC006981.1 | - | 4, 5 | 449 | |||
BC066945.1 | - | 6, 7 | 711 | 4837780 | ||
BM979591.1 | - | 1, 2 | 105 | |||
BX647100.1 | - | 4, 5 | 399 | |||
EU667388.1 | - | 4, 5 | 408 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
