siRNA Details
Gene Symbol: LARP7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001267039.1 | NP_001253968.1 | 9 | 1456 | ||
NM_015454.2 | NP_056269.1 | 7 | 1120 | |||
NM_016648.3 | NP_057732.2 | 7 | 1248 | |||
NR_049768.1 | 7 | 1145 | ||||
GenBank mRNA | AK000274.1 | - | 7 | 1058 | ||
AK225900.1 | - | 4 | 617 | |||
AL049996.1 | - | 1 | 95 | |||
BC006981.1 | - | 7 | 1032 | |||
BC066945.1 | - | 9 | 1294 | 4837780 | ||
BC107709.1 | - | 1 | 89 | |||
BX647100.1 | - | 7 | 982 | |||
EU667388.1 | - | 7 | 991 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
