siRNA Details
Gene Symbol: ZNF248 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001267605.1 | NP_001254534.1 | 1 | 432 | ||
NM_001267606.1 | NP_001254535.1 | 1 | 432 | |||
NM_001267607.1 | NP_001254536.1 | 1 | 432 | |||
NM_021045.2 | NP_066383.1 | 1 | 432 | |||
XM_006717485.3 | XP_006717548.1 | 1 | 487 | |||
XM_017016445.1 | XP_016871934.1 | 1 | 432 | |||
XM_017016449.1 | XP_016871938.1 | 1 | 476 | |||
XM_017016450.1 | XP_016871939.1 | 1 | 472 | |||
XM_017016451.1 | XP_016871940.1 | 1 | 432 | |||
GenBank mRNA | AJ491695.1 | - | 1 | 354 | ||
AJ492196.1 | - | 1 | 354 | |||
BC021819.1 | - | 1 | 347 | |||
BC110869.1 | - | 1 | 222 | |||
BC132915.1 | - | 1 | 90 | 40146937 | ||
BC132917.1 | - | 1 | 90 | 40146939 | ||
DB073240.1 | - | 1 | 432 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
