siRNA Details
Gene Symbol: ZNF248 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001267597.1 | NP_001254526.1 | 3 | 205 | ||
NM_001267605.1 | NP_001254534.1 | 3 | 577 | |||
NM_001267606.1 | NP_001254535.1 | 1 | 1326 | |||
NM_001267607.1 | NP_001254536.1 | 3 | 577 | |||
NM_021045.2 | NP_066383.1 | 3 | 577 | |||
XM_006717485.3 | XP_006717548.1 | 2 | 534 | |||
XM_011519597.2 | XP_011517899.1 | 2 | 126 | |||
XM_011519598.2 | XP_011517900.1 | 3 | 227 | |||
XM_011519599.1 | XP_011517901.1 | 3 | 180 | |||
XM_011519600.2 | XP_011517902.1 | 2 | 68 | |||
XM_017016445.1 | XP_016871934.1 | 2 | 674 | |||
XM_017016446.1 | XP_016871935.1 | 3 | 225 | |||
XM_017016449.1 | XP_016871938.1 | 2 | 523 | |||
XM_017016450.1 | XP_016871939.1 | 2 | 519 | |||
XM_017016451.1 | XP_016871940.1 | 2 | 674 | |||
GenBank mRNA | AJ491695.1 | - | 3 | 499 | ||
AJ492196.1 | - | 3 | 499 | |||
AK299357.1 | - | 3 | 205 | |||
BC021819.1 | - | 2 | 394 | |||
BC110869.1 | - | 1 | 1116 | |||
BC132915.1 | - | 3 | 235 | 40146937 | ||
BC132917.1 | - | 3 | 235 | 40146939 | ||
BC146820.1 | - | 2 | 74 | |||
DA128760.1 | - | 2 | 138 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
