siRNA Details
Gene Symbol: ARL10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317948.1 | NP_001304877.1 | 1 | 83 | ||
NM_173664.5 | NP_775935.1 | 1 | 83 | |||
XM_011534529.2 | XP_011532831.1 | 1 | 77 | |||
XM_011534530.2 | XP_011532832.1 | 1 | 77 | |||
XM_011534531.2 | XP_011532833.1 | 1 | 77 | |||
XM_017009372.1 | XP_016864861.1 | 1 | 77 | |||
GenBank mRNA | BC059361.1 | - | 1 | 83 | 30344341 | |
DR001833.1 | - | 1 | 57 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
