siRNA Details
Gene Symbol: CYB5RL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001031672.2 | NP_001026842.2 | 6 | 793 | ||
GenBank mRNA | AK090456.1 | - | 6 | 2904 | ||
BC012781.2 | - | 5 | 577 | |||
BC049198.1 | - | 4 | 407 | |||
BC064419.1 | - | 6 | 766 | |||
CD515193.1 | - | 4 | 539 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
