siRNA Details
Gene Symbol: CYB5RL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001031672.2 | NP_001026842.2 | 3 | 445 | ||
GenBank mRNA | AK090456.1 | - | 3 | 383 | ||
BC012781.2 | - | 3 | 378 | |||
BC049198.1 | - | 1 | 102 | |||
BC064419.1 | - | 3 | 408 | |||
BC071735.1 | - | 3 | 384 | |||
CD515193.1 | - | 1 | 191 | |||
DA793739.1 | - | 3 | 445 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
