siRNA Details
Gene Symbol: RPLP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004.3 | NP_000995.1 | 1 | 60 | ||
GenBank mRNA | AK130959.1 | - | 1 | 28 | ||
AK311954.1 | - | 1 | 28 | |||
BC005354.1 | - | 1 | 31 | 4052568 | ||
BC005920.1 | - | 1 | 29 | 4274135 | ||
CN270352.1 | - | 1 | 66 | |||
D28411.1 | - | 1 | 29 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
