siRNA Details
Gene Symbol: ATP8B5P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003581.2 | 11 | 935 | |||
NR_003582.1 | 11 | 939 | ||||
NR_110320.1 | 11 | 935 | ||||
GenBank mRNA | AK310278.1 | - | 11 | 911 | ||
BC031276.1 | - | 11 | 922 | |||
BC130448.1 | - | 6 | 699 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
