siRNA Details
Gene Symbol: KLHL7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001031710.2 | NP_001026880.2 | 1 | 370 | ||
NM_001172428.1 | NP_001165899.1 | 1 | 370 | |||
NR_033328.1 | 1 | 370 | ||||
NR_033329.1 | 1 | 370 | ||||
XM_006715753.2 | XP_006715816.1 | 1 | 365 | |||
XM_006715754.2 | XP_006715817.1 | 1 | 317 | |||
XM_006715757.3 | XP_006715820.1 | 1 | 365 | |||
XM_017012440.1 | XP_016867929.1 | 1 | 365 | |||
XM_017012441.1 | XP_016867930.1 | 1 | 317 | |||
GenBank mRNA | AF111113.1 | - | 1 | 254 | ||
AK056390.1 | - | 1 | 321 | |||
AK290479.1 | - | 1 | 164 | |||
AK296219.1 | - | 1 | 295 | |||
AK297369.1 | - | 295 | ||||
AK297421.1 | - | 1 | 371 | |||
AK297595.1 | - | 1 | 314 | |||
AL136597.1 | - | 1 | 316 | |||
BC009555.1 | - | 1 | 286 | |||
BC039585.1 | - | 1 | 312 | 5527346 | ||
DC370850.1 | - | 1 | 370 | |||
EF560731.1 | - | 1 | 89 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
