siRNA Details
Gene Symbol: RPS15A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001019.4 | NP_001010.2 | 1 | 21 | ||
GenBank mRNA | AK291988.1 | - | 1 | 21 | ||
BF678907.1 | - | 1 | 22 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
