siRNA Details
Gene Symbol: NSUN5P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK094312.1 | - | 1 | 69 | ||
AK126486.1 | - | 3 | 2043 | |||
AK128374.1 | - | 1 | 281 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
