siRNA Details
Gene Symbol: LINC01547 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027128.1 | 1 | 90 | |||
NR_027129.1 | 1 | 90 | ||||
GenBank mRNA | AF380178.1 | - | 1 | 22 | ||
AK127059.1 | - | 1 | 90 | |||
AY040088.1 | - | 1 | 22 | |||
BI829356.1 | - | 1 | 54 | |||
BI907880.1 | - | 1 | 90 |
Gene Symbol: FAM207A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BG744869.1 | - | 6 | 663 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
