siRNA Details
Gene Symbol: IFITM3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021034.2 | NP_066362.2 | 1 | 79 | ||
NR_049759.1 | 1 | 79 | ||||
GenBank mRNA | AK292173.1 | - | 1 | 23 | ||
BC006794.1 | - | 1 | 37 | 2986145 | ||
BC008417.1 | - | 1 | 22 | 4075453 | ||
BC022439.1 | - | 1 | 28 | 4282809 | ||
BC070243.1 | - | 1 | 13 | 30401041 | ||
DB261103.1 | - | 1 | 79 | |||
X57352.1 | - | 1 | 215 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
