siRNA Details
Gene Symbol: NXF5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032946.2 | NP_116564.2 | 3 | 275 | ||
NR_028089.1 | 3 | 275 | ||||
GenBank mRNA | AJ277654.1 | - | 3 | 275 | ||
AJ277655.1 | - | 3 | 275 | |||
AJ277656.1 | - | 3 | 275 | |||
AJ277657.1 | - | 3 | 275 | |||
AJ277658.1 | - | 3 | 275 | |||
BC131708.1 | - | 2 | 96 | 40121743 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
