siRNA Details
Gene Symbol: TCP11X2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277423.1 | NP_001264352.1 | 1 | 12 | ||
GenBank mRNA | AK093310.1 | - | 1 | 12 | ||
AK307893.1 | - | 1 | 12 |
Gene Symbol: NXF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK303069.1 | - | 1 | 42 |
Gene Symbol: TCP11X1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017030010.1 | XP_016885499.1 | 1 | 36 | ||
XM_017030011.1 | XP_016885500.1 | 1 | 34 | |||
XM_017030014.1 | XP_016885503.1 | 1 | 12 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
