siRNA Details
Gene Symbol: NBPF12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278141.1 | NP_001265070.1 | 2 | 661 | ||
GenBank mRNA | AB033071.1 | - | 1 | 214 |
Gene Symbol: NBPF11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_183372.5 | NP_899228.4 | 2 | 664 | ||
NR_046188.3 | 2 | 664 | ||||
NR_110864.1 | 2 | 634 | ||||
GenBank mRNA | AL832622.2 | - | 2 | 797 | ||
BC136292.1 | - | 2 | 664 | 9020279 | ||
BM802549.1 | - | 1 | 89 | |||
BP873295.1 | - | 2 | 500 | |||
BX648497.1 | - | 2 | 433 | |||
DB062330.1 | - | 3 | 402 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
