siRNA Details
Gene Symbol: NBPF11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_183372.5 | NP_899228.4 | 1 | 129 | ||
NR_046188.3 | 1 | 129 | ||||
NR_110864.1 | 1 | 129 | ||||
GenBank mRNA | AL832622.2 | - | 1 | 262 | ||
BC136292.1 | - | 1 | 129 | 9020279 | ||
BI552657.1 | - | 1 | 245 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
