siRNA Details
Gene Symbol: LCN6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198946.2 | NP_945184.1 | 1 | 126 | ||
GenBank mRNA | AF303084.1 | - | 1 | 123 | ||
AY301269.1 | - | 1 | 81 | |||
AY358598.1 | - | 1 | 106 | |||
BC062746.1 | - | 1 | 127 | 6452008 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
