siRNA Details
Gene Symbol: CXorf40A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001171908.2 | NP_001165379.1 | 1, 2 | 327 | ||
NM_001324274.1 | NP_001311203.1 | 1, 2 | 178 | |||
NM_001324275.1 | NP_001311204.1 | 1, 2 | 178 | |||
NM_001324277.1 | NP_001311206.1 | 1, 2 | 860 | |||
NM_001324278.1 | NP_001311207.1 | 1, 2 | 327 | |||
NM_001324279.1 | NP_001311208.1 | 1, 2 | 327 | |||
NM_001324280.1 | NP_001311209.1 | 1, 2 | 860 | |||
GenBank mRNA | AK055235.1 | - | 1 | 1385 | ||
AY074889.1 | - | 1, 2 | 340 | |||
BC143662.1 | - | 1, 2 | 175 | 9052176 | ||
BG105508.1 | - | 203 | ||||
BI819316.1 | - | 1, 2 | 71 | |||
DC339764.1 | - | 1, 2 | 86 | |||
U66049.1 | - | 1, 2 | 318 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
