siRNA Details
Gene Symbol: CXorf40A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001171907.2 | NP_001165378.1 | 1 | 97 | ||
NM_001171908.2 | NP_001165379.1 | 1 | 97 | |||
NM_001324276.1 | NP_001311205.1 | 1 | 97 | |||
NM_001324277.1 | NP_001311206.1 | 1 | 97 | |||
NM_001324278.1 | NP_001311207.1 | 1 | 97 | |||
NM_001324279.1 | NP_001311208.1 | 1 | 97 | |||
NM_001324280.1 | NP_001311209.1 | 1 | 97 | |||
NM_178124.5 | NP_835225.2 | 1 | 97 | |||
GenBank mRNA | AK055235.1 | - | 1 | 429 | ||
AK291899.1 | - | 1 | 23 | |||
AK294153.1 | - | 1 | 73 | |||
AY074889.1 | - | 1 | 59 | |||
BC065539.1 | - | 1 | 22 | 6143134 | ||
BE265359.1 | - | 1 | 34 | |||
CB156531.1 | - | 1 | 97 | |||
U66049.1 | - | 1 | 88 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
